6-(Difluoromethyl)pyridin-3-amine 97%

Lieferant: Apollo Scientific

PC201370-250MG PC201370-1G
APOSPC201370-250MGEA 683 EUR
APOSPC201370-250MG APOSPC201370-1G
6-(Difluoromethyl)pyridin-3-amine 97%
Formel: C₆H₆F₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18384489
CAS-Nummer: 913090-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR