3-Chloro-4-fluoro-5-(trifluoromethyl)aniline 98%

Lieferant: Apollo Scientific

PC201372-250MG PC201372-1G PC201372-5G
APOSPC201372-250MGEA 63.8 EUR
APOSPC201372-250MG APOSPC201372-1G APOSPC201372-5G
3-Chloro-4-fluoro-5-(trifluoromethyl)aniline 98%
3-Chloro-4-fluoro-5-(trifluoromethyl)aniline
Formel: C₇H₄ClF₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD09033155
CAS-Nummer: 914225-61-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR