5-Bromo-4-fluoropyridin-2-amine 95%

Lieferant: Apollo Scientific
Achtung

PC201380-5G
APOSPC201380-5GEA 105 EUR
APOSPC201380-5G
5-Bromo-4-fluoropyridin-2-amine 95%
5-Bromo-4-fluoropyridin-2-amine
Formel: C₅H₄BrFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16658678
CAS-Nummer: 944401-69-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR