7-Amino-5-fluoroindolin-2-one 95+%

Lieferant: Apollo Scientific

PC201382-250MG
APOSPC201382-250MGEA 42.2 EUR
APOSPC201382-250MG
7-Amino-5-fluoroindolin-2-one 95+%
7-Amino-5-fluoroindolin-2-one
Formel: C₈H₇FN₂O
Molecular Weight: 166.152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13193359
CAS-Nummer: 945381-62-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR