6-Chloro-5-(trifluoromethyl)pyridin-3-amine 95+%

Lieferant: Apollo Scientific

PC201388-100MG
APOSPC201388-100MGEA 0 EUR
APOSPC201388-100MG
6-Chloro-5-(trifluoromethyl)pyridin-3-amine 95+%
6-Chloro-5-(trifluoromethyl)pyridin-3-amine
Formel: C₆H₄ClF₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11101042
CAS-Nummer: 99368-68-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR