5-(4-Chlorophenyl)-2-(trifluoromethyl)furan-3-carboxylic acid 97%

Lieferant: Apollo Scientific

Synonyme: 5-(4-Chlorophenyl)-2-(trifluoromethyl)-3-furoic acid

PC2022-1G
APOSPC2022-1GEA 87.3 EUR
APOSPC2022-1G
5-(4-Chlorophenyl)-2-(trifluoromethyl)furan-3-carboxylic acid 97%
5-(4-Chlorophenyl)-2-(trifluoromethyl)furan-3-carboxylic acid
Formel: C₁₂H₆ClF₃O₃
Molecular Weight: 290,63 g/mol
Schmelzpunkt: 188…190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00275555
CAS-Nummer: 175276-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR