2-Amino-5-(trifluormethoxy)benzoesäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-amino-5-(trifluoromethoxy)benzoic acid

PC2033-1G
APOSPC2033-1GEA 21.9 EUR
APOSPC2033-1G
2-Amino-5-(trifluormethoxy)benzoesäure 98%
2-Amino-5-(trifluoromethoxy)benzoic acid
Formel: C₈H₆F₃NO₃
Molecular Weight: 221,14 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00176651
CAS-Nummer: 83265-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR