7-Fluoro-4-hydroxyquinoline
Lieferant: Apollo Scientific
Synonyme:
7-Fluoro-4-hydroxy-1-azanaphthalene, 7-Fluoroquinolin-4-ol
PC2037-1G
APOSPC2037-1GEA
375
EUR
APOSPC2037-1G
7-Fluoro-4-hydroxyquinoline
7-Fluoro-4-hydroxyquinoline
Formel:
C₉H₆FNO Molecular Weight: 163,148 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269612 CAS-Nummer: 391-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...