2-Amino-5,6,7,8-tetrahydro-4-(trifluoromethyl)quinazoline 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Trifluoromethyl)-5,6,7,8-tetrahydroquinazolin-2-amine

PC2048-1G
APOSPC2048-1GEA 263 EUR
APOSPC2048-1G
2-Amino-5,6,7,8-tetrahydro-4-(trifluoromethyl)quinazoline 97%
2-Amino-5,6,7,8-tetrahydro-4-(trifluoromethyl)quinazoline
Formel: C₉H₁₀F₃N₃
Molecular Weight: 217.191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01312182
CAS-Nummer: 256954-38-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR