2-Amino-3,3,3-trifluoropropionic acid hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,3,3-Trifluoro-DL-alanine hydrochloride

PC2052-1G
APOSPC2052-1GEA 253 EUR
APOSPC2052-1G
2-Amino-3,3,3-trifluoropropionic acid hydrochloride 97%
2-Amino-3,3,3-trifluoropropionic acid hydrochloride
Formel: C₃H₅ClF₃NO₂
Molecular Weight: 179,53 g/mol
Schmelzpunkt: 186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04973034
CAS-Nummer: 96105-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR