4-Chloro-6-(trifluoromethyl)-1H-benzimidazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2073R-1G
PC2073R-500MG
PC2073R-5G
APOSPC2073R-1GEA
0
EUR
APOSPC2073R-1G
APOSPC2073R-500MG
APOSPC2073R-5G
4-Chloro-6-(trifluoromethyl)-1H-benzimidazole 97%
4-Chloro-6-(trifluoromethyl)-1H-benzimidazole
Formel:
C₈H₄ClF₃N₂ Schmelzpunkt: 210…212 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067733 CAS-Nummer: 175135-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...