4-[2-Chloro-5-(trifluoromethyl)phenyl]-3-thiosemicarbazide 97%

Lieferant: Apollo Scientific

PC2074R-1G
APOSPC2074R-1GEA 26.3 EUR
APOSPC2074R-1G
4-[2-Chloro-5-(trifluoromethyl)phenyl]-3-thiosemicarbazide 97%
4-[2-Chloro-5-(trifluoromethyl)phenyl]-3-thiosemicarbazide
Formel: C₈H₇ClF₃N₃S
Schmelzpunkt: 162…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041274
CAS-Nummer: 206559-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR