4-Chlor-7-(trifluormethyl)chinolin 98%

Lieferant: Apollo Scientific

PC2075M-1G PC2075M-5G
APOSPC2075M-1GEA 21.9 EUR
APOSPC2075M-1G APOSPC2075M-5G
4-Chlor-7-(trifluormethyl)chinolin 98%
4-Chloro-7-(trifluoromethyl)quinoline
Formel: C₁₀H₅ClF₃N
Schmelzpunkt: 69…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006775
CAS-Nummer: 346-55-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR