Copper(II) trifluoroacetylacetonate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Copper(ii)trifluoroacetylacetonate

PC2096-5G
APOSPC2096-5GEA 74.1 EUR
APOSPC2096-5G
Copper(II) trifluoroacetylacetonate 97%
Copper(II) trifluoroacetylacetonate
Schmelzpunkt: 198…199 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042511
CAS-Nummer: 14324-82-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR