2-Chloro-6-(trifluoromethyl)-1,3-benzoxazole

Lieferant: Apollo Scientific
Achtung

PC210005-1G
APOSPC210005-1GEA 423 EUR
APOSPC210005-1G
2-Chloro-6-(trifluoromethyl)-1,3-benzoxazole
2-Chloro-6-(trifluoromethyl)-1,3-benzoxazole
Formel: C₈H₃ClF₃NO
Lagertemperatur: Kühlschrank
MDL: MFCD18250485
CAS-Nummer: 1248393-79-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR