1-(4-Nitrophenyl)-4-(trifluoromethyl)-1H-pyrazole
Lieferant: Apollo Scientific
PC210006-1G
APOSPC210006-1GEA
562
EUR
APOSPC210006-1G
1-(4-Nitrophenyl)-4-(trifluoromethyl)-1H-pyrazole
1-(4-Nitrophenyl)-4-(trifluoromethyl)-1H-pyrazole
Formel:
C₁₀H₆F₃N₃O₂ Schmelzpunkt: 93…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD28023542 CAS-Nummer: 1393125-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...