2-(Acetylamino)-3-(3,5-difluorophenyl)-2-propenoic acid

Lieferant: Apollo Scientific

PC210007-1G
APOSPC210007-1GEA 321 EUR
APOSPC210007-1G
2-(Acetylamino)-3-(3,5-difluorophenyl)-2-propenoic acid
2-(Acetylamino)-3-(3,5-difluorophenyl)-2-propenoic acid
Formel: C₁₁H₉F₂NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD27680350
CAS-Nummer: 883035-24-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR