Methyl ester-N-(6,8-difluoro-2H-1-benzopyran-3-yl)carbamic acid

Lieferant: Apollo Scientific

PC210008-500MG
APOSPC210008-500MGEA 894 EUR
APOSPC210008-500MG
Methyl ester-N-(6,8-difluoro-2H-1-benzopyran-3-yl)carbamic acid
Methyl ester-N-(6,8-difluoro-2H-1-benzopyran-3-yl)carbamic acid
Formel: C₁₁H₉F₂NO₃
Molecular Weight: 241,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20528372
CAS-Nummer: 1034001-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR