tert-Butyl (3R,4S)-3-fluoro-4-hydroxypiperidine-1-carboxylate ≥ ee 98%

Lieferant: Apollo Scientific

Synonyme: (3R,4S)-tert-Butyl 3-fluoro-4-hydroxypiperidine-1-carboxylate

PC210010-500MG
APOSPC210010-500MGEA 214 EUR
APOSPC210010-500MG
tert-Butyl (3R,4S)-3-fluoro-4-hydroxypiperidine-1-carboxylate ≥ ee 98%
tert-Butyl (3R,4S)-3-fluoro-4-hydroxypiperidine-1-carboxylate
Formel: C₁₀H₁₈FNO₃
Molecular Weight: 219,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18633012
CAS-Nummer: 1174020-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR