(3S,4R)-tert-Butyl-3-fluoro-4-hydroxypiperidine-1-carboxylate ≥ ee 98%

Lieferant: Apollo Scientific

Synonyme: (3S,4R)-tert-Butyl 3-fluoro-4-hydroxypiperidine-1-carboxylate

PC210011-500MG
APOSPC210011-500MGEA 336 EUR
APOSPC210011-500MG
(3S,4R)-tert-Butyl-3-fluoro-4-hydroxypiperidine-1-carboxylate ≥ ee 98%
(3S,4R)-tert-Butyl-3-fluoro-4-hydroxypiperidine-1-carboxylate
Formel: C₁₀H₁₈FNO₃
Molecular Weight: 219,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18632748
CAS-Nummer: 1174020-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR