4-Fluorphenylboronsäureneopentylglycolester

Lieferant: Apollo Scientific

Synonyme: 2-(4-Fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane

PC210017-5G
APOSPC210017-5GEA 59.6 EUR
APOSPC210017-5G
4-Fluorphenylboronsäureneopentylglycolester
4-Fluorphenylboronsäureneopentylglycolester
Formel: C₁₁H₁₄BFO₂
Molecular Weight: 208.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05663850
CAS-Nummer: 225916-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR