2-(2-Bromo-5-fluorophenoxy)-3-(dimethylamino)prop-2-enenitrile
Lieferant: Apollo Scientific
PC210034-500MG
APOSPC210034-500MGEA
560
EUR
APOSPC210034-500MG
2-(2-Bromo-5-fluorophenoxy)-3-(dimethylamino)prop-2-enenitrile
2-(2-Bromo-5-fluorophenoxy)-3-(dimethylamino)prop-2-enenitrile
Formel:
C₁₁H₁₀BrFN₂O Molecular Weight: 285,112 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28101529 CAS-Nummer: 1620478-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...