5-(2-Bromo-5-fluorophenoxy)pyrimidine-2,4-diamine

Lieferant: Apollo Scientific

PC210035-500MG
APOSPC210035-500MGEA 0 EUR
APOSPC210035-500MG
5-(2-Bromo-5-fluorophenoxy)pyrimidine-2,4-diamine
5-(2-Bromo-5-fluorophenoxy)pyrimidine-2,4-diamine
Formel: C₁₀H₈BrFN₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD09261189
CAS-Nummer: 935534-20-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR