N-(4-Bromo-2-methoxyphenyl)-2,2,2-trifluoroacetamide

Lieferant: Apollo Scientific

PC210037-500MG
APOSPC210037-500MGEA 782 EUR
APOSPC210037-500MG
N-(4-Bromo-2-methoxyphenyl)-2,2,2-trifluoroacetamide
N-(4-Bromo-2-methoxyphenyl)-2,2,2-trifluoroacetamide
Formel: C₉H₇BrF₃NO₂
Molecular Weight: 298,057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28101534
CAS-Nummer: 870838-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR