2-(Dimethylamino)-4-fluoro-3-formylbenzonitrile

Lieferant: Apollo Scientific

PC210041-500MG
APOSPC210041-500MGEA 1350 EUR
APOSPC210041-500MG
2-(Dimethylamino)-4-fluoro-3-formylbenzonitrile
2-(Dimethylamino)-4-fluoro-3-formylbenzonitrile
Formel: C₁₀H₉FN₂O
Molecular Weight: 192,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28101537
CAS-Nummer: 1515395-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR