4-Bromo-1-(4-fluorophenyl)butan-1-one
Lieferant: Apollo Scientific
PC210044-1G
PC210044-500MG
APOSPC210044-1GEA
894
EUR
APOSPC210044-1G
APOSPC210044-500MG
4-Bromo-1-(4-fluorophenyl)butan-1-one
4-Bromo-1-(4-fluorophenyl)butan-1-one
Formel:
C₁₀H₁₀BrFO Molecular Weight: 245.088 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16323231 CAS-Nummer: 40132-01-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...