(2R,4S)-4-[(tert-Butyl)(diphenyl)silyloxy]-2-fluorocyclohexan-1-one

Lieferant: Apollo Scientific

PC210046-500MG
APOSPC210046-500MGEA 782 EUR
APOSPC210046-500MG
(2R,4S)-4-[(tert-Butyl)(diphenyl)silyloxy]-2-fluorocyclohexan-1-one
(2R,4S)-4-[(tert-Butyl)(diphenyl)silyloxy]-2-fluorocyclohexan-1-one
Formel: C₂₂H₂₇FO₂Si
Molecular Weight: 370,532 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28101544
CAS-Nummer: 1268512-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR