2-Amino-1-[4-(2-fluorophenyl)piperazin-1-yl]ethan-1-one hydrochloride
Lieferant: Apollo Scientific
PC210051-1G
APOSPC210051-1GEA
780
EUR
APOSPC210051-1G
2-Amino-1-[4-(2-fluorophenyl)piperazin-1-yl]ethan-1-one hydrochloride
2-Amino-1-[4-(2-fluorophenyl)piperazin-1-yl]ethan-1-one hydrochloride
Formel:
C₁₂H₁₆FN₃O·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD28041366 CAS-Nummer: 1605772-63-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...