(Z)-2-[2-(4-Fluorophenyl)-4-oxo-4H,5H-pyrazolo[1,5-a]pyrazin-5-yl]-N'-hydroxyethenimidamide
Lieferant: Apollo Scientific
PC210052-1G
APOSPC210052-1GEA
780
EUR
APOSPC210052-1G
(Z)-2-[2-(4-Fluorophenyl)-4-oxo-4H,5H-pyrazolo[1,5-a]pyrazin-5-yl]-N'-hydroxyethenimidamide
(Z)-2-[2-(4-Fluorophenyl)-4-oxo-4H,5H-pyrazolo[1,5-a]pyrazin-5-yl]-N'-hydroxyethenimidamide
Formel:
C₁₄H₁₂FN₅O₂ Molecular Weight: 301.276 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17780265 CAS-Nummer: 1255791-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...