Ethyl 4-amino-2-sulphanylidene-3-[3-(trifluoromethyl)phenyl]-2,3-dihydro-1,3-thiazole-5-carboxylate

Lieferant: Apollo Scientific

PC210054-1G
APOSPC210054-1GEA 780 EUR
APOSPC210054-1G
Ethyl 4-amino-2-sulphanylidene-3-[3-(trifluoromethyl)phenyl]-2,3-dihydro-1,3-thiazole-5-carboxylate
Ethyl 4-amino-2-sulphanylidene-3-[3-(trifluoromethyl)phenyl]-2,3-dihydro-1,3-thiazole-5-carboxylate
Formel: C₁₃H₁₁F₃N₂O₂S₂
Molecular Weight: 348.364 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02089347
CAS-Nummer: 312922-28-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR