6-(Trifluoromethyl)-[1,2]oxazolo[4,5-b]pyridin-3-amine

Lieferant: Apollo Scientific

PC210056-500MG
APOSPC210056-500MGEA 539 EUR
APOSPC210056-500MG
6-(Trifluoromethyl)-[1,2]oxazolo[4,5-b]pyridin-3-amine
6-(Trifluoromethyl)-[1,2]oxazolo[4,5-b]pyridin-3-amine
Formel: C₇H₄F₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD28122584
CAS-Nummer: 1622069-57-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR