3-Chloro-2-(2-methyl(8-quinolyloxy))-5-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
PC210057-1G
APOSPC210057-1GEA
241
EUR
APOSPC210057-1G
3-Chloro-2-(2-methyl(8-quinolyloxy))-5-(trifluoromethyl)pyridine
3-Chloro-2-(2-methyl(8-quinolyloxy))-5-(trifluoromethyl)pyridine
Formel:
C₁₆H₁₀ClF₃N₂O Molecular Weight: 338.712 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00170103 CAS-Nummer: 1024367-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...