N'-Hydroxy-2-{[4-(trifluoromethoxy)phenyl]sulphonyl}ethanimidamide

Lieferant: Apollo Scientific

PC210058-1G
APOSPC210058-1GEA 139 EUR
APOSPC210058-1G
N'-Hydroxy-2-{[4-(trifluoromethoxy)phenyl]sulphonyl}ethanimidamide
N'-Hydroxy-2-{[4-(trifluoromethoxy)phenyl]sulphonyl}ethanimidamide
Formel: C₉H₉F₃N₂O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD09757711
CAS-Nummer: 884504-68-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR