Methyl-2-(2-nitrilo-3-fluorophenylthio)acetate

Lieferant: Apollo Scientific

Synonyme: Methyl 2-[(2-cyano-3-fluorophenyl)sulphanyl]acetate

PC210060-1G
APOSPC210060-1GEA 139 EUR
APOSPC210060-1G
Methyl-2-(2-nitrilo-3-fluorophenylthio)acetate
Methyl-2-(2-nitrilo-3-fluorophenylthio)acetate
Formel: C₁₀H₈FNO₂S
Molecular Weight: 225,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00171056
CAS-Nummer: 262433-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR