2-(2-(N-(2,4-Difluorophenyl)carbamoyl)phenyl)acetic acid

Lieferant: Apollo Scientific

Synonyme: 2-{2-[(2,4-Difluoroanilino)carbonyl]phenyl}acetic acid

PC210061-1G
APOSPC210061-1GEA 139 EUR
APOSPC210061-1G
2-(2-(N-(2,4-Difluorophenyl)carbamoyl)phenyl)acetic acid
2-(2-(N-(2,4-Difluorophenyl)carbamoyl)phenyl)acetic acid
Formel: C₁₅H₁₁F₂NO₃
Molecular Weight: 291,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00171195
CAS-Nummer: 811841-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR