Methyl-4-(2-nitro-4-(trifluoromethyl)phenoxy)benzoate
Lieferant: Apollo Scientific
Synonyme:
Methyl 4-[2-nitro-4-(trifluoromethyl)phenoxy]benzenecarboxylate
PC210062-1G
APOSPC210062-1GEA
139
EUR
APOSPC210062-1G
Methyl-4-(2-nitro-4-(trifluoromethyl)phenoxy)benzoate
Methyl-4-(2-nitro-4-(trifluoromethyl)phenoxy)benzoate
Formel:
C₁₅H₁₀F₃NO₅ Molecular Weight: 341,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00245361 CAS-Nummer: 297148-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...