N-(2,4-Difluorophenyl)-4,4-dimethyl-3-oxopentanamide

Lieferant: Apollo Scientific

PC210064-1G
APOSPC210064-1GEA 139 EUR
APOSPC210064-1G
N-(2,4-Difluorophenyl)-4,4-dimethyl-3-oxopentanamide
N-(2,4-Difluorophenyl)-4,4-dimethyl-3-oxopentanamide
Formel: C₁₃H₁₅F₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01567378
CAS-Nummer: 497059-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR