Methyl (2S)-2-acetamido-3-(2,3,6-trifluorophenyl)propanoate ≥ ee 80%
Lieferant: Apollo Scientific
PC210068-500MG
APOSPC210068-500MGEA
519
EUR
APOSPC210068-500MG
Methyl (2S)-2-acetamido-3-(2,3,6-trifluorophenyl)propanoate ≥ ee 80%
Methyl (2S)-2-acetamido-3-(2,3,6-trifluorophenyl)propanoate
Formel:
C₁₂H₁₂F₃NO₃ Molecular Weight: 275,224 g/mol Schmelzpunkt: 110…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD28127033 CAS-Nummer: 1630114-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...