Methyl 2-acetamido-3-(2,3,6-trifluorophenyl)propanoate
Lieferant: Apollo Scientific
PC210070-500MG
APOSPC210070-500MGEA
1260
EUR
APOSPC210070-500MG
Methyl 2-acetamido-3-(2,3,6-trifluorophenyl)propanoate
Methyl 2-acetamido-3-(2,3,6-trifluorophenyl)propanoate
Formel:
C₁₂H₁₂F₃NO₃ Molecular Weight: 275,224 g/mol Schmelzpunkt: 115…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD28127036 CAS-Nummer: 1630096-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...