3-chloro-N'-hydroxy-5-(trifluoromethyl)-2-pyridinecarboximidamide

Lieferant: Apollo Scientific

PC210085-500MG
APOSPC210085-500MGEA 536 EUR
APOSPC210085-500MG
3-chloro-N'-hydroxy-5-(trifluoromethyl)-2-pyridinecarboximidamide
3-chloro-N'-hydroxy-5-(trifluoromethyl)-2-pyridinecarboximidamide
Formel: C₇H₅ClF₃N₃O
Molecular Weight: 239,582 g/mol
Schmelzpunkt: 127…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00231716
CAS-Nummer: 303997-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR