(2-amino-4,5-difluorophenyl)methanol

Lieferant: Apollo Scientific

PC210091-10G
APOSPC210091-10GEA 1260 EUR
APOSPC210091-10G
(2-amino-4,5-difluorophenyl)methanol
(2-amino-4,5-difluorophenyl)methanol
Formel: C₇H₇F₂NO
Molecular Weight: 159,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18825440
CAS-Nummer: 748805-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR