3-chloro-2-{[(2Z)-2-(nitroimino)imidazolidin-1-yl]methyl}-5-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

PC210095-500MG
APOSPC210095-500MGEA 749 EUR
APOSPC210095-500MG
3-chloro-2-{[(2Z)-2-(nitroimino)imidazolidin-1-yl]methyl}-5-(trifluoromethyl)pyridine
3-chloro-2-{[(2Z)-2-(nitroimino)imidazolidin-1-yl]methyl}-5-(trifluoromethyl)pyridine
Formel: C₁₀H₉ClF₃N₅O₂
Molecular Weight: 323,659 g/mol
Schmelzpunkt: 162…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD28805992
CAS-Nummer: 1781254-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR