tert-butyl 3-[(4-fluorophenyl)methyl]-4-oxo-3H,4H,5H,6H,7H,8H-pyrido[4,3-d]pyrimidine-6-carboxylate

Lieferant: Apollo Scientific

PC210096-500MG
APOSPC210096-500MGEA 1070 EUR
APOSPC210096-500MG
tert-butyl 3-[(4-fluorophenyl)methyl]-4-oxo-3H,4H,5H,6H,7H,8H-pyrido[4,3-d]pyrimidine-6-carboxylate
tert-butyl 3-[(4-fluorophenyl)methyl]-4-oxo-3H,4H,5H,6H,7H,8H-pyrido[4,3-d]pyrimidine-6-carboxylate
Formel: C₁₉H₂₂FN₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28805993
CAS-Nummer: 1781241-50-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR