8-chloro-3-phenyl-6-(trifluoromethyl)imidazo[1,5-a]pyridine
Lieferant: Apollo Scientific
PC210097-500MG
APOSPC210097-500MGEA
640
EUR
APOSPC210097-500MG
8-chloro-3-phenyl-6-(trifluoromethyl)imidazo[1,5-a]pyridine
8-chloro-3-phenyl-6-(trifluoromethyl)imidazo[1,5-a]pyridine
Formel:
C₁₄H₈ClF₃N₂ Molecular Weight: 296,675 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28805997 CAS-Nummer: 1781241-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...