1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(propan-2-yl)piperazine

Lieferant: Apollo Scientific

PC210098-500MG
APOSPC210098-500MGEA 1070 EUR
APOSPC210098-500MG
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(propan-2-yl)piperazine
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(propan-2-yl)piperazine
Formel: C₁₃H₁₇ClF₃N₃
Molecular Weight: 307,742 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28806000
CAS-Nummer: 1781241-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR