[6-(trifluoromethyl)pyridin-3-yl]methyl 6-(trifluoromethyl)pyridine-3-carboxylate
Lieferant: Apollo Scientific
PC210099-500MG
APOSPC210099-500MGEA
857
EUR
APOSPC210099-500MG
[6-(trifluoromethyl)pyridin-3-yl]methyl 6-(trifluoromethyl)pyridine-3-carboxylate
[6-(trifluoromethyl)pyridin-3-yl]methyl 6-(trifluoromethyl)pyridine-3-carboxylate
Formel:
C₁₄H₈F₆N₂O₂ Schmelzpunkt: 102…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD28806001 CAS-Nummer: 1781241-48-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...