tert-butyl 4-{[3-(4-fluorophenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl}piperazine-1-carboxylate

Lieferant: Apollo Scientific

PC210100-500MG
APOSPC210100-500MGEA 749 EUR
APOSPC210100-500MG
tert-butyl 4-{[3-(4-fluorophenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl}piperazine-1-carboxylate
tert-butyl 4-{[3-(4-fluorophenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl}piperazine-1-carboxylate
Formel: C₁₉H₂₆FN₃O₄
Molecular Weight: 379,426 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28806004
CAS-Nummer: 1781241-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR