tert-butyl N-(1-{[3-(4-fluorophenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl}piperidin-3-yl)carbamate

Lieferant: Apollo Scientific

PC210101-500MG
APOSPC210101-500MGEA 749 EUR
APOSPC210101-500MG
tert-butyl N-(1-{[3-(4-fluorophenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl}piperidin-3-yl)carbamate
tert-butyl N-(1-{[3-(4-fluorophenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl}piperidin-3-yl)carbamate
Formel: C₂₀H₂₈FN₃O₄
Molecular Weight: 393,452 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28806006
CAS-Nummer: 1781241-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR