3-bromo-4-chloro-5-fluoropyridine hydrochloride

Lieferant: Apollo Scientific

PC210103-500MG
APOSPC210103-500MGEA 640 EUR
APOSPC210103-500MG
3-bromo-4-chloro-5-fluoropyridine hydrochloride
3-bromo-4-chloro-5-fluoropyridine hydrochloride
Formel: C₅H₂BrClFN·ClH
Molecular Weight: 246,892 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28805788
CAS-Nummer: 1211540-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR