potassium (3-ethoxyphenyl)trifluoroboranuide

Lieferant: Apollo Scientific

PC210107-1G
APOSPC210107-1GEA 139 EUR
APOSPC210107-1G
potassium (3-ethoxyphenyl)trifluoroboranuide
potassium (3-ethoxyphenyl)trifluoroboranuide
Formel: C₈H₉BF₃O·K
Molecular Weight: 228,061 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09993311
CAS-Nummer: 1638533-83-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR